Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxylesterase 1/CES1 Antibody (SR1990), Novus Biologicals™
SDP

Catalog No. NB258763 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258763 50 μg
NB258138 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258763 Supplier Novus Biologicals Supplier No. NBP32194050UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carboxylesterase 1/CES1 Monoclonal antibody specifically detects Carboxylesterase 1/CES1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation

Specifications

Antigen Carboxylesterase 1/CES1
Applications Western Blot, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone SR1990
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Flow Cytometry 1:100, Immunoprecipitation 1:40
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias ACAT, acyl coenzyme A:cholesterol acyltransferase, Acyl-coenzyme A:cholesterol acyltransferase, Brain carboxylesterase hBr1, carboxylesterase 1, carboxylesterase 1 (monocyte/macrophage serine esterase 1), carboxylesterase 2 (liver), CEH, Ces-1, CES1A2, CES2EC 3.1.1.1, cholesteryl ester hydrolase, Cocaine carboxylesterase, EC 3.1.1, egasyn, HMSE1, HMSECES1A1, human monocyte/macrophage serine esterase 1, liver carboxylesterase 1, MGC117365, Monocyte/macrophage serine esterase, PCE-1, REH, Retinyl ester hydrolase, Serine esterase 1, Ses-1, SES1TGH, Triacylglycerol hydrolase
Host Species Rabbit
Immunogen A synthesized peptide derived from human Carboxylesterase 1/CES1 (Uniprot #: P23141)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, CD Markers, Endocrinology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1066
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.