Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxylesterase 1C/CES1C Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Carboxylesterase 1C/CES1C |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Carboxylesterase 1C/CES1C Polyclonal specifically detects Carboxylesterase 1C/CES1C in Mouse samples. It is validated for Western Blot.Specifications
Carboxylesterase 1C/CES1C | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
ES1 | |
The immunogen is a synthetic peptide directed towards the C region of mouse ES1 (NP_031980.2). Peptide sequence TNLSKMVMKFWANFARNGNPNGEGLPHWPEYDEQEGYLQIGATTQQAQRL | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
13884 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title