Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxylesterase 2/CES2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215864
Description
Carboxylesterase 2/CES2 Polyclonal specifically detects Carboxylesterase 2/CES2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CES2 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
carboxylesterase 2 (intestine, liver), carboxylesterase 2EC 3.1.1.1, CE-2PCE-2, CES2A1, EC 3.1.1, EC 3.1.1.84, hCE-2, iCE, intestinal carboxylesterase; liver carboxylesterase-2 | |
Rabbit | |
69 kDa | |
0.1 mL | |
Lipid and Metabolism | |
8824 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
O00748 | |
CES2 | |
Recombinant protein encompassing a sequence within the C-terminus region of human CES2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction