Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxylesterase 3/CES3/Esterase 31 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299711AF700
Description
Carboxylesterase 3/CES3/Esterase 31 Polyclonal specifically detects Carboxylesterase 3/CES3/Esterase 31 in Human samples. It is validated for ELISA.Specifications
Carboxylesterase 3/CES3/Esterase 31 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
23491 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 700 | |
carboxylesterase 3, carboxylesterase 3 (brain), EC 3.1.1, EC 3.1.1.1, ES31FLJ21736, esterase 31, Liver carboxylesterase 31 homolog | |
Produced in rabbits immunized with purified, recombinant Carboxylesterase 3/CES3/Esterase 31 (NP_079198.2; Met 1-Glu 561). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction