Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxylesterase 3/CES3/Esterase 31 Rabbit anti-Human, Clone: 372, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | Carboxylesterase 3/CES3/Esterase 31 |
---|---|
Clone | 372 |
Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
Carboxylesterase 3/CES3/Esterase 31 Monoclonal specifically detects Carboxylesterase 3/CES3/Esterase 31 in Human samples. It is validated for Western Blot, ELISA.Specifications
Carboxylesterase 3/CES3/Esterase 31 | |
Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
23491 | |
IgG | |
Protein A purified |
372 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Human | |
carboxylesterase 3, carboxylesterase 3 (brain), EC 3.1.1, EC 3.1.1.1, ES31FLJ21736, esterase 31, Liver carboxylesterase 31 homolog | |
Recombinant Human Carboxylesterase 3/CES3/Esterase 31 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title