Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxymethylcellulose Sodium, Low Viscosity, USP, Spectrum™ Chemical
SDP

Catalog No. 18601476
Click to view available options
Quantity:
100 g
125 g
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail

CA193, 9004-32-4

Spectrum™ Chemical Carboxymethylcellulose Sodium, Low Viscosity, Viscosity of 2 Percent Aqueous Solution @ 25 DEG C: 37.5 cP, USP is often used as a thickener and stabilizer. The USP grade indicates it is a grade suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-32-4
Molecular Formula (C12 H14 O9 R6)n
Molecular Weight (g/mol) 263.20
MDL Number MFCD00081472
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O

Specifications

CAS Max % 100%
Quantity 125 g
Grade USP
Loss on Drying 10%
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.