Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase A1/CPA1 Antibody (CPA1/2712), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279880H
Description
Carboxypeptidase A1/CPA1 Monoclonal specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Carboxypeptidase A1/CPA1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CPA1 | |
Recombinant full-length human Carboxypeptidase A1/CPA1 protein (Uniprot: P15085) | |
0.1 mL | |
Mast Cell Markers | |
Human | |
IgG2c κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
CPA1/2712 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction