Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase A1/CPA1 Antibody (rCPA1/8688), Novus Biologicals™

Mouse Monoclonal Antibody
$216.50 - $443.50
Specifications
Antigen | Carboxypeptidase A1/CPA1 |
---|---|
Clone | rCPA1/8688 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
Carboxypeptidase A1/CPA1 Monoclonal antibody specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Carboxypeptidase A1/CPA1 | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Mast Cell Markers | |
10mM PBS with 0.05% BSA | |
1357 | |
IgG1 κ | |
Protein A or G purified |
rCPA1/8688 | |
Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1 | |
Recombinant full-length human Carboxypeptidase A1/CPA1 protein | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title