Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carboxypeptidase A1 Monoclonal Antibody (CL6607)
GREENER_CHOICE

Catalog No. PIMA531440
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531440 Supplier Invitrogen™ Supplier No. MA531440
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: DFVGHQVLRI SVADEAQVQK VKELEDLEHL QLDFWRGPAH PGSPIDVRVP FPSIQAVKIF LESHGISYET Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 77%.

CPA1, also known as CPA, is a 419 amino acid secreted monomeric protein that is highly expressed in pancreatic tissue. Functioning as a pancreatic exopeptidase, CPA1 uses zinc as a cofactor to catalyze the release of C-terminal amino acids from a variety of proteins, thereby playing a key role in protein digestion and degradation. Via its catalytic activity, CPA1 is also thought to be involved in zymogen (proenzyme) inhibition, probably functioning to block enzyme activation pathways. Abnormal levels of CPA1 are associated with pancreatic cancer, suggesting a possible role in either tumor progression or tumor suppression events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxypeptidase A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL6607
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cpa1
Gene Accession No. P15085
Gene Alias 0910001L12Rik; carboxypeptidase A; carboxypeptidase A1; carboxypeptidase A1 (pancreatic); carboxypeptidase A1, pancreatic; CPA; Cpa1; pancreatic carboxypeptidase A; pancreatic procarboxypeptidase A
Gene Symbols Cpa1
Host Species Mouse
Immunogen Recombinant protein fragment of Human CPA1 (Product #RP-92630)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1357
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.