Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase A2/CPA2 Antibody (009), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289530F
Description
Carboxypeptidase A2/CPA2 Monoclonal specifically detects Carboxypeptidase A2/CPA2 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
Carboxypeptidase A2/CPA2 | |
Monoclonal | |
FITC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
1358 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
9 | |
Sandwich ELISA Detection | |
carboxypeptidase A2, carboxypeptidase A2 (pancreatic), EC 3.4.17, EC 3.4.17.15 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Carboxypeptidase A2/CPA2 (NP_001859.1; Met1-Tyr417). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction