Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase A2/CPA2 Antibody (179), DyLight 755, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290717IR
Description
Carboxypeptidase A2/CPA2 Monoclonal specifically detects Carboxypeptidase A2/CPA2 in Mouse samples. It is validated for ELISA.Specifications
Carboxypeptidase A2/CPA2 | |
Monoclonal | |
DyLight 755 | |
carboxypeptidase A2, carboxypeptidase A2 (pancreatic), EC 3.4.17, EC 3.4.17.15 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carboxypeptidase A2/CPA2 (Q504N0; Met1-Tyr417). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
179 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
1358 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction