Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase A2/CPA2 Antibody (OTI3D8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270443H
Description
Carboxypeptidase A2/CPA2 Monoclonal specifically detects Carboxypeptidase A2/CPA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Carboxypeptidase A2/CPA2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CPA2 | |
Full length recombinant protein of human Carboxypeptidase A2/CPA2 (NP_001860) produced in HEK293T cells. | |
0.1 mL | |
1358.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3D8 | |
Western Blot, Immunohistochemistry | |
carboxypeptidase A2, carboxypeptidase A2 (pancreatic), EC 3.4.17, EC 3.4.17.15 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction