Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carboxypeptidase A2 Monoclonal Antibody (1G7G10E4)
GREENER_CHOICE

Catalog No. PIMA529180
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529180 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529180 Supplier Invitrogen™ Supplier No. MA529180
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CPA2/Carboxypeptidase A2.

Three different forms of human pancreatic procarboxypeptidase A have been isolated. The encoded protein represents the A2 form, which is a monomeric protein with different biochemical properties from the A1 and A3 forms. The A2 form of pancreatic procarboxypeptidase acts on aromatic C-terminal residues and is a secreted protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxypeptidase A2
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1G7G10E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CPA2
Gene Accession No. P48052
Gene Alias carboxypeptidase A2; carboxypeptidase A2 (pancreatic); carboxypeptidase A2, pancreatic; CBPA2; Cpa2; EC 3.4.17; EC 3.4.17.15; LOW QUALITY PROTEIN: carboxypeptidase A2; zgc:92530
Gene Symbols CPA2
Host Species Mouse
Immunogen Recombinant Human CPA2 protein (Met1-Tyr417).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1358
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.