Learn More
Invitrogen™ Carboxypeptidase A2 Monoclonal Antibody (384023)

Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CPA1, rhCPA4, rhCPB1, or rhCPE is observed. This antibody recognizes the pro form (aa 17-417), but not the activated enzyme (aa 113-417) in Western blots. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | Carboxypeptidase A2 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 384023 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CPA2 |
| Gene Accession No. | P48052 |
| Gene Alias | carboxypeptidase A2; carboxypeptidase A2 (pancreatic); carboxypeptidase A2, pancreatic; CBPA2; Cpa2; EC 3.4.17; EC 3.4.17.15; LOW QUALITY PROTEIN: carboxypeptidase A2; zgc:92530 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.