Learn More
Invitrogen™ Carboxypeptidase A2 Polyclonal Antibody

Description
The antibody detects endogenous levels of total Carboxypeptidase A2 / CPA2 protein.
Specifications
Specifications
| Antigen | Carboxypeptidase A2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CPA2 |
| Gene Accession No. | P48052, Q504N0 |
| Gene Alias | carboxypeptidase A2; carboxypeptidase A2 (pancreatic); carboxypeptidase A2, pancreatic; CBPA2; Cpa2; EC 3.4.17; EC 3.4.17.15; LOW QUALITY PROTEIN: carboxypeptidase A2; zgc:92530 |
| Gene Symbols | CPA2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.