Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carboxypeptidase A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589787
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589787 Supplier Invitrogen™ Supplier No. PA589787
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGNFNFGAYH TLEEISQEMD NLVAEHPGLV SKVNIGSSFE NRPMNVLKFS TGGDKPAIWL DAGIHAREWV TQATALWTAN KIVSDYGKDP SITSILDALD IFLLPVTNPD GYVFSQTKNR MWRKTRSKVS GSLCVGVDPN RNWDAGFGGP GASSNPCSDS YHGPSANSEV EVKSIVDFIK SHGKVKAFIT LHSYSQLLMF PYGYKCTKLD DFDELSEVAQ KAAQSLRSLH GTKYKVGPIC SVIYQASGGS IDWSYDYGIK YSFAFELRDT GRYGFLLPAR QILPTAEETW LGLKAIMEHV RDHPY; Positive Samples: U-87MG, SH-SY5Y, Mouse pancreas, Rat pancreas.

Three different forms of human pancreatic procarboxypeptidase A have been isolated. The encoded protein represents the A2 form, which is a monomeric protein with different biochemical properties from the A1 and A3 forms. The A2 form of pancreatic procarboxypeptidase acts on aromatic C-terminal residues and is a secreted protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxypeptidase A2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CPA2
Gene Accession No. P19222, P48052, Q504N0
Gene Alias carboxypeptidase A2; carboxypeptidase A2 (pancreatic); carboxypeptidase A2, pancreatic; CBPA2; Cpa2; EC 3.4.17; EC 3.4.17.15; LOW QUALITY PROTEIN: carboxypeptidase A2; zgc:92530
Gene Symbols CPA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 115-419 of human CPA2 (NP_0018602).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1358, 232680, 296959
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.