Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carboxypeptidase A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531333
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531333 Supplier Invitrogen™ Supplier No. PA531333
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HepG2. Predicted reactivity: Mouse (83%), Rat (84%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Three different forms of human pancreatic procarboxypeptidase A have been isolated. This gene encodes a form which is obtained as a binary complex of a procarboxypeptidase A with proproteinase E and functions as a secretory granule metalloexopeptidase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxypeptidase A3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CPA3
Gene Accession No. P15088
Gene Alias Carboxypeptidase A3; carboxypeptidase A3 (mast cell); carboxypeptidase A3, mast cell; Cpa3; Mast cell carboxypeptidase A; MC-CPA; mMC-CPA; R-CPA; RMC-CP; tissue carboxypeptidase A
Gene Symbols CPA3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CPA3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1359
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.