Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Carboxypeptidase A5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570412
Description
This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Rat: 93%.
Carboxypeptidases have functions ranging from digestion of food to selective biosynthesis of neuroendocrine peptides. Members of the A/B subfamily of carboxypeptidases, such as CPA5, contain an approximately 90-amino acid pro region that assists in the folding of the active carboxypeptidase domain. Cleavage of the pro region activates the enzyme.
Specifications
Carboxypeptidase A5 | |
Polyclonal | |
Unconjugated | |
CPA5 | |
4930430M09Rik; carboxypeptidase A5; CPA5; testicular tissue protein Li 32 | |
Rabbit | |
Affinity chromatography | |
RUO | |
408212 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
CPA5 | |
synthetic peptide directed towards the middle region of human Carboxypeptidase A5. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction