Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase B1/CPB1 Antibody (108), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289532MFV450
Description
Carboxypeptidase B1/CPB1 Monoclonal antibody specifically detects Carboxypeptidase B1/CPB1 in Human samples. It is validated for ELISASpecifications
Carboxypeptidase B1/CPB1 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
1360 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
108 | |
Sandwich ELISA Detection | |
carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, DKFZp779K1333, EC 3.4.17, EC 3.4.17.2, Pancreas-specific protein, PASP, PCPB, procarboxypeptidase B | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Carboxypeptidase B1/CPB1 (Accession#: NP_001862.2; Met1-Tyr417). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction