Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase B1/CPB1 Antibody (117), Alexa Fluor™ 750

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290555AF750
Description
Carboxypeptidase B1/CPB1 Monoclonal specifically detects Carboxypeptidase B1/CPB1 in Mouse samples. It is validated for ELISA.Specifications
Carboxypeptidase B1/CPB1 | |
Monoclonal | |
Alexa Fluor 750 | |
carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, DKFZp779K1333, EC 3.4.17, EC 3.4.17.2, Pancreas-specific protein, PASP, PCPB, procarboxypeptidase B | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carboxypeptidase B1/CPB1 (NP_083982.1; Met 1-Tyr 415). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Human CPB1 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
117 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
1360 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction