Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carboxypeptidase B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547416
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547416 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547416 Supplier Invitrogen™ Supplier No. PA547416
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, approximately 10% cross-reactivity with recombinant human (rh) CPB1 is observed and less than 1% cross-reactivity with rhCPA1 and recombinant mouse CPA4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxypeptidase B1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CPB1
Gene Accession No. 0
Gene Alias 0910001A18Rik; 1810063F02Rik; 2210008M23Rik; AI504870; carboxypeptidase B; carboxypeptidase B1; carboxypeptidase B1 (tissue); Cpb; Cpb1; Pancreas-specific protein; pancreatic carboxypeptidase B; pancreatic carboxypeptidase B1; PASP; PCPB; procarboxypeptidase B; protaminase; tissue carboxypeptidase B
Gene Symbols CPB1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Carboxypeptidase B1/CPB1 Ser16-Tyr415.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76703
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.