Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase B2/CPB2 Antibody (OTI3D5), Janelia Fluor™ 646, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270444JF646

Catalog No. N270444J646


Only null left
Add to Cart

Description

Description

Carboxypeptidase B2/CPB2 Monoclonal specifically detects Carboxypeptidase B2/CPB2 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Carboxypeptidase B2/CPB2
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CPB2
Recombinant protein fragment corresponding to amino acids 224-423 of human Carboxypeptidase B2/CPB2 (NP_001863) produced in E.coli.
0.1 mL
Primary
Human
IgG1
Immunohistochemistry
OTI3D5
Immunohistochemistry
carboxypeptidase B2, carboxypeptidase B2 (plasma), carboxypeptidase B2 (plasma, carboxypeptidase U), Carboxypeptidase U, CPUEC 3.4.17.20, EC 3.4.17, PCPB, Plasma carboxypeptidase B, TAFIcarboxypeptidase B-like protein, Thrombin-activable fibrinolysis inhibitor, thrombin-activatable fibrinolysis inhibitor
Mouse
Affinity Purified
Lipid and Metabolism
1361.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.