Learn More
Invitrogen™ Carboxypeptidase B2 Monoclonal Antibody (5026)

Description
MA1-43024 detects TAFI in human samples. MA1-43024 has successfully been used in WB, ELISA, IA applications. MA1-43024 was derived from human TAFI and activated TAFI.
Specifications
Specifications
| Antigen | Carboxypeptidase B2 |
| Applications | ELISA, Inhibition Assays, Western Blot |
| Classification | Monoclonal |
| Clone | 5026 |
| Concentration | 10.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50% water with 50% glycerol and no preservative |
| Gene | CPB2 |
| Gene Accession No. | Q96IY4 |
| Gene Alias | 1110032P04Rik; 4930405E17Rik; AI255929; carboxypeptidase B2; carboxypeptidase B2 (plasma); carboxypeptidase B2 (plasma, carboxypeptidase U); carboxypeptidase B-like protein; carboxypeptidase R; Carboxypeptidase U; Cpb2; CPR; CPU; pCPB; Plasma carboxypeptidase B; Tafi; thrombin-activable fibrinolysis inhibitor; thrombin-activatable fibrinolysis inhibitor |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.