Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carboxypeptidase B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579063
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579063 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579063 Supplier Invitrogen™ Supplier No. PA579063
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, HEPG2 whole cell. IHC: human lung cancer tissue.

Carboxypeptidases are enzymes that hydrolyze C-terminal peptide bonds. The carboxypeptidase family includes metallo-, serine, and cysteine carboxypeptidases. According to their substrate specificity, these enzymes are referred to as carboxypeptidase A (cleaving aliphatic residues) or carboxypeptidase B (cleaving basic amino residues). CPB2 is activated by trypsin and acts on carboxypeptidase B substrates. After thrombin activation, the mature protein downregulates fibrinolysis. Polymorphisms have been described for its gene and its promoter region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxypeptidase B2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene CPB2
Gene Accession No. Q96IY4
Gene Alias 1110032P04Rik; 4930405E17Rik; AI255929; carboxypeptidase B2; carboxypeptidase B2 (plasma); carboxypeptidase B2 (plasma, carboxypeptidase U); carboxypeptidase B-like protein; carboxypeptidase R; Carboxypeptidase U; Cpb2; CPR; CPU; pCPB; Plasma carboxypeptidase B; Tafi; thrombin-activable fibrinolysis inhibitor; thrombin-activatable fibrinolysis inhibitor
Gene Symbols CPB2
Host Species Rabbit
Immunogen E. coli-derived human CPB2 recombinant protein (Position: K166-D388).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1361
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.