Learn More
Invitrogen™ Carboxypeptidase B2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110680
Description
Immunogen sequence: QVLAALPRTS RQVQVLQNLT TTYEIVLWQP VTADLIVKKK QVHFFVNASD VDNVKAHLNV SGIPCSVLLA DVEDLIQQQI SNDTVSPRAS ASYYEQYHSL NEIYSWIEFI TERHPDMLTK IHIGSSFEKY PLYVLKVSGK EQAA Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 81%.
Carboxypeptidases are enzymes that hydrolyze C-terminal peptide bonds. The carboxypeptidase family includes metallo-, serine, and cysteine carboxypeptidases. According to their substrate specificity, these enzymes are referred to as carboxypeptidase A (cleaving aliphatic residues) or carboxypeptidase B (cleaving basic amino residues). CPB2 is activated by trypsin and acts on carboxypeptidase B substrates. After thrombin activation, the mature protein downregulates fibrinolysis. Polymorphisms have been described for its gene and its promoter region.
Specifications
| Carboxypeptidase B2 | |
| Polyclonal | |
| Unconjugated | |
| CPB2 | |
| 1110032P04Rik; 4930405E17Rik; AI255929; carboxypeptidase B2; carboxypeptidase B2 (plasma); carboxypeptidase B2 (plasma, carboxypeptidase U); carboxypeptidase B-like protein; carboxypeptidase R; Carboxypeptidase U; Cpb2; CPR; CPU; pCPB; Plasma carboxypeptidase B; Tafi; thrombin-activable fibrinolysis inhibitor; thrombin-activatable fibrinolysis inhibitor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1361 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96IY4 | |
| CPB2 | |
| Recombinant Protein corresonding to Human Carboxypeptidase B2. Recombinant protein control fragment (Product #RP-102322). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.