Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carboxypeptidase D Polyclonal Antibody

Catalog No. PIPA5111417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111417 Supplier Invitrogen™ Supplier No. PA5111417

Rabbit Polyclonal Antibody

Immunogen sequence: KIGQTNARGK DLDTDFTNNA SQPETKAIIE NLIQKQDFSL SVALDGGSML VTYPYDKPVQ TVENKETLKH LASLYANNHP SMHMGQPSCP NKSD.

The metallocarboxypeptidase family of enzymes is divided into 2 subfamilies based on sequence similarities. The pancreatic carboxypeptidase-like and the regulatory B-type carboxypeptidase subfamilies. Carboxypeptidase D has been identified as a regulatory B-type carboxypeptidase. CPD is a homolog of duck gp180, a hepatitis B virus-binding protein. Transcript variants utilizing alternative polyadenylation signals exist for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxypeptidase D
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CPD
Gene Accession No. O75976
Gene Alias AA960140; AW322530; carboxypeptidase D; CPD; cpd {ECO:0000312; D830034L15Rik; gp180; metallocarboxypeptidase D; RGD:2393}
Gene Symbols CPD
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Carboxypeptidase D. Recombinant protein control fragment (Product #RP-106636).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1362
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.