Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase E/CPE Antibody (1), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289258G
Description
Carboxypeptidase E/CPE Monoclonal antibody specifically detects Carboxypeptidase E/CPE in Human samples. It is validated for ELISASpecifications
Carboxypeptidase E/CPE | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
1 | |
ELISA | |
carboxypeptidase E, Carboxypeptidase H, CPH, EC 3.4.17, EC 3.4.17.10, Enkephalin convertase, Prohormone-processing carboxypeptidase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Carboxypeptidase E/CPE (NP_001864.1; Met 1-Ser 453). | |
0.1 mL | |
Neuroscience | |
1363 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction