Learn More
Carboxypeptidase E/CPE Antibody, DyLight 650, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP298739C
Catalog No. NB082839
Only null left
Add to Cart
Description
Description
Carboxypeptidase E/CPE Polyclonal specifically detects Carboxypeptidase E/CPE in Human samples. It is validated for ELISA.
Specifications
Specifications
Carboxypeptidase E/CPE
Polyclonal
50 mM Sodium Borate
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA
DyLight 650
carboxypeptidase E, Carboxypeptidase H, CPH, EC 3.4.17, EC 3.4.17.10, Enkephalin convertase, Prohormone-processing carboxypeptidase
Produced in rabbits immunized with purified, recombinant Carboxypeptidase E/CPE (NP_001864.1; Met 1-Ser 453).
0.1 mL
Neuroscience
1363
Store at 4°C in the dark.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.