Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase E/CPE Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298739MFV450
Description
Carboxypeptidase E/CPE Polyclonal antibody specifically detects Carboxypeptidase E/CPE in Human samples. It is validated for ELISASpecifications
Carboxypeptidase E/CPE | |
Polyclonal | |
ELISA | |
carboxypeptidase E, Carboxypeptidase H, CPH, EC 3.4.17, EC 3.4.17.10, Enkephalin convertase, Prohormone-processing carboxypeptidase | |
Produced in rabbits immunized with purified, recombinant Human Carboxypeptidase E/CPE (Accession#: NP_001864.1; Met 1-Ser 453) | |
0.1 mL | |
Neuroscience | |
1363 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction