Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Carboxypeptidase E Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577027
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a carboxypeptidase that cleaves C-terminal amino acid residues and is involved in the biosynthesis of peptide hormones and neurotransmitters, including insulin. It is a peripheral membrane protein. The protein specifically binds regulated secretory pathway proteins, including prohormones, but not constitutively secreted proteins. Mutations in this gene are implicated in type II diabetes.
Specifications
Carboxypeptidase E | |
Polyclonal | |
Unconjugated | |
CPE | |
CARBE; Carboxypeptidase E; carboxypeptidase H; CarboxypeptidaseE; CarboxypeptidaseH; CBPE; cobalt-stimulated chromaffin granule carboxypeptidase; CPE; CPH; Cph 1; Cph1; Cph-1; enkephalin convertase; fat; insulin granule-associated carboxypeptidase; prohormone-processing carboxypeptidase; R74677; si:ch211-198g9.2; unnamed protein product; zgc:85981 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12876, 1363, 25669 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.2 | |
P15087, P16870, Q00493 | |
CPE | |
Recombinant full length Human Carboxypeptidase E. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction