Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase M Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298372B
Description
Carboxypeptidase M Polyclonal specifically detects Carboxypeptidase M in Mouse samples. It is validated for ELISA.Specifications
Carboxypeptidase M | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
1368 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Biotin | |
carboxypeptidase M, EC 3.4.17, EC 3.4.17.12 | |
Produced in rabbits immunized with purified, recombinant Carboxypeptidase M (Q80V42-1; Met1-His422). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction