Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase M Antibody (OTI2D11), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270446V
Description
Carboxypeptidase M Monoclonal specifically detects Carboxypeptidase M in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Carboxypeptidase M | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CPM | |
Recombinant protein fragment corresponding to amino acids 124-423 of human Carboxypeptidase M (NP_001865) produced in E.coli. | |
0.1 mL | |
1368.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2D11 | |
Western Blot, Immunohistochemistry | |
carboxypeptidase M, EC 3.4.17, EC 3.4.17.12 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction