Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carboxypeptidase M Monoclonal Antibody (OTI4E11)
GREENER_CHOICE

Catalog No. PIMA526863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526863 Supplier Invitrogen™ Supplier No. MA526863
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Carboxypeptidase M Monoclonal Antibody for Western Blot, IHC (P)

Specifically removes C-terminal basic residues (Arg or Lys) from peptides and proteins. It is believed to play important roles in the control of peptide hormone and growth factor activity at the cell surface, and in the membrane-localized degradation of extracellular proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxypeptidase M
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CPM
Gene Accession No. P14384
Gene Alias 1110060I01Rik; 5730456K23Rik; AA589379; Carboxypeptidase M; Cpm; E030045M14Rik; renal carboxypeptidase; urinary carboxypeptidase B
Gene Symbols CPM
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 124-423 of CPM produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1368
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.