Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

carboxypeptidase M, Mouse anti-Rat, Clone: 7F9, Millipore Sigma™

Catalog No. MABS192525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS192525 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS192525 Supplier MilliporeSigma Supplier No. MABS192525UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-carboxypeptidase M, clone 7F9, Cat. No. MABS1925, is a mouse monoclonal antibody that detects Carboxypeptidase M and has been tested for use in Immunofluorescence, Immunohistochemistry, Immunoprecipitation, and Western Blotting.

Carboxypeptidase M (UniProt: D4A9Q5; also known as CPM) is encoded by the Cpm gene (Gene ID: 314855) in rat. Rat CPM a glycosylphosphatidylinositol (GPI) anchored metallo-carboxypeptidase that cleaves basic carboxyl-terminal amino acids from various peptides at neutral pH. Rat CPM is synthesized with a signal peptide (aa 1-17), which is subsequently cleaved off to generate the mature form. The mature CPM displays high specificity and cleaves only C-terminal arginine or lysine residues in a variety of peptide substrates. It is shown to be expressed in the lung and placenta at high levels. Its expression is detected in alveolar cells lacking T1alpha, a type I cell marker protein found in adult lung. Carboxypeptidase M also serves as a macrophage differentiation marker, which is preferentially induced in epithelioid cells of all granuloma types studied, but not in resting macrophages. Hence, it can be employed for detection of minute granuloma even in the presence of non-granulomatous macrophages. Clone 7F9 recognizes CPM expressed in late embryonic rat lung to postnatal day 1. (Ref.: Fujiwara, N., et al. (2007), Respirology 12(1); 54-62; Tsakiris, I., et al. (2012). Lab. Invest. 92(3); 345-361).
TRUSTED_SUSTAINABILITY

Specifications

Antigen carboxypeptidase M
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7F9
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected carboxypeptidase M in Western Blotting applications (Fujiwara, N., et. al. (2007). Respirology. 12(1):54-62).Immunoprecipitation Analysis: A representative lot immunoprecipitated carboxypeptidase M in Immunoprecipitation applications (Fujiwara, N., et. al. (2007). Respirology. 12(1):54-62).Immunohistochemistry Analysis: A representative lot detected carboxypeptidase M in Immunohistochemistry applications (Fujiwara, N., et. al. (2007). Respirology. 12(1):54-62).
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias CPM;CP-M
Gene Symbols Cpm
Host Species Mouse
Immunogen Membrane fraction of E18.5 embryonic rat lung.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001101568.1
Test Specificity Clone 7F9 specifically detects carboxypeptidase M in rat tissues.
Target Species Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.