Learn More
Invitrogen™ Carboxypeptidase M Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593064
Description
Immunogen sequence: VQATGALYSR SLTPDDDVFQ YLAHTYASRN PNMKKGDECK NKMNFPNGVT NGYSWYPLQG GMQDYNYIWA QCFEITLELS CCKYPREEKL PSFWNNNKAS LIEYIKQVHL GVKGQVFDQN GNPLPNVIVE VQDRKHICPY RTNKYGEYYL LLLPGSYIIN VTVPGHDPHI TKVIIPEKSQ NFSALKKDIL LPFQGQLDSI PVSNPSCPMI PLYRNLPDHS AATKPSLFLF LVSLLHIFFK; Positive Samples: HeLa; Cellular Location: Cell membrane, GPI-anchor, Lipid-anchor.
Specifically removes C-terminal basic residues (Arg or Lys) from peptides and proteins. It is believed to play important roles in the control of peptide hormone and growth factor activity at the cell surface, and in the membrane-localized degradation of extracellular proteins.
Specifications
Carboxypeptidase M | |
Polyclonal | |
Unconjugated | |
CPM | |
1110060I01Rik; 5730456K23Rik; AA589379; Carboxypeptidase M; Cpm; E030045M14Rik; renal carboxypeptidase; urinary carboxypeptidase B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1368, 314855 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P14384 | |
CPM | |
Recombinant fusion protein containing a sequence corresponding to amino acids 204-443 of human CPM (NP_0010055021). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.