Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase Y Mouse anti-Yeast, Clone: 10A5B5, Abnova™

Catalog No. 89035427 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-427 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-427 Supplier Abnova Corporation Supplier No. MAB14022

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against yeast Carboxypeptidase Y.

  • High specificity
  • High reactivity

Specifications

Antigen Carboxypeptidase Y
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 10A5B5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against yeast Carboxypeptidase Y.
Dilution Immunocytochemistry (20 ug/mL) Immunofluorescence (20 ug/mL) Western Blot (0.25 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In HEPES (0.02% sodium azide).
Host Species Mouse
Immunogen Purified yeast Carboxypeptidase Y.
Purification Method Ig fraction
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.