Learn More
Carboxypeptidase Y Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | Carboxypeptidase Y |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0803
![]() ![]() |
Rockland Immunochemicals
2004011350100 |
100 μg |
Each for $636.24
|
|
|||||
50-199-0804
![]() ![]() |
Rockland Immunochemicals
200401135S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-CARBOXYPEPTIDASE Y is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum as well as purified and partially purified Carboxypeptidase Y [Baker's Yeast]. Cross reactivity against Carboxypeptidase Y from other tissues and species may occur but have not been specifically determined.
Carboxypeptidase Y is Involved in the degradation of small peptides. It digests peptides containing an aliphatic or hydrophobic residue in P1' position, as well as methionine, leucine or phenylalanine in the P1 position of an ester substrate.Specifications
Carboxypeptidase Y | |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Yeast | |
Carboxypeptidase Y; cpy1; pcy1 | |
Primary | |
Antibody |
1 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
Carboxypeptidase Y [Baker's Yeast] | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.