Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carcinoembryonic Antigen Mouse anti-Human, Clone: SPM330, Abnova™

Catalog No. 89933955 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-955 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-955 Supplier Abnova Corporation Supplier No. MAB13043
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native human Carcinoembryonic Antigen.

Specifications

Antigen Carcinoembryonic Antigen
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SPM330
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human Carcinoembryonic Antigen.
Dilution Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS.
Host Species Mouse
Immunogen Native purified human Carcinoembryonic Antigen.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with nonspecific cross-reacting antigen (NCA) and shows a cross-reaction with human polymorphonuclear leukocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues.
Target Species Human
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.