Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carcinoma Glycans Mouse anti-Human, Unconjugated, Clone: mAb-A4, MilliporeSigma™

Catalog No. MABC17092MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC17092MI 25 μg
MABC17091MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC17092MI Supplier MilliporeSigma Supplier No. MABC170925UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Carcinoma Glycans Monoclonal antibody specifically detects Carcinoma Glycans in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carcinoma Glycans
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone mAb-A4
Conjugate Unconjugated
Gene Alias Cancer Cell Glycans
Host Species Mouse
Immunogen Live HES-3 human embryonic stem cells.
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone mAb-A4 detects ovarian and breast cancer cells and targets H Type 1 glycans.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.