Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARD11/CARMA1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299265APC
Description
CARD11/CARMA1 Polyclonal antibody specifically detects CARD11/CARMA1 in Mouse samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
CARD11/CARMA1 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
BIMP3card-maguk protein 1, CARD-containing MAGUK protein 1, Carma 1, CARMA1bcl10-interacting maguk protein 3, caspase recruitment domain family, member 11, caspase recruitment domain-containing protein 11, MGC133069 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CARD11/CARMA1 | |
0.1 mL | |
Apoptosis | |
84433 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction