Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARD12 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen CARD12
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:1000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018188
SDP
View Documents
Novus Biologicals
NB500225SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB500225
SDP
View Documents
Novus Biologicals
NB500225
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

CARD12 Polyclonal specifically detects CARD12 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CARD12
Western Blot 1:500-1:1000
Polyclonal
Rabbit
Apoptosis, Immunology, Innate Immunity
PBS with 0.02% Sodium Azide
58484
N terminus (MNFIKDNSRALIQRMGM) of the A and B isoforms of the human CLAN protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1 mg/mL
Western Blot
Unconjugated
RUO
Human
CARD12leucine rich repeat and CARD domaincontaining 4, caspase recruitment domain family, member 12, Caspase recruitment domain-containing protein 12, Clan protein, CLAN1ipaf, CLANCARD, LRR, and NACHT-containing protein, CLR2.1, Ice protease-activating factor, Ipaf, NLR family, CARD domain containing 4
NLRC4
IgG
Affinity Purified
Reacts with the N terminus (MNFIKDNSRALIQRMGM) of the A and B isoforms of the human CLAN protein.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.