Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARD12 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$243.50 - $390.00

Specifications

Antigen CARD12
Dilution Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin 1:1000 - 1:5000, Immunohistochemistry-Frozen 1:10 - 1:500
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056142T
SDP
View Documents
Novus Biologicals
NB10056142SS
0.025 mL
Each for $243.50
Only null left
Add to Cart
 
NB10056142
SDP
View Documents
Novus Biologicals
NB10056142
0.05 mL
Each for $390.00
Only null left
Add to Cart
 
Description

Description

CARD12 Polyclonal specifically detects CARD12 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

CARD12
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Rabbit
Apoptosis, Immunology, Innate Immunity
Whole antisera with 0.05% Sodium Azide
CARD12leucine rich repeat and CARD domaincontaining 4, caspase recruitment domain family, member 12, Caspase recruitment domain-containing protein 12, Clan protein, CLAN1ipaf, CLANCARD, LRR, and NACHT-containing protein, CLR2.1, Ice protease-activating factor, Ipaf, NLR family, CARD domain containing 4
NLRC4
IgG
Unpurified
Ipaf
Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin 1:1000 - 1:5000, Immunohistochemistry-Frozen 1:10 - 1:500
Polyclonal
Antisera
RUO
Human, Mouse
Q9NPP4
58484
A synthetic peptide corresponding to within amino acis 610 and 660 of Ipaf/Clan/CARD12 was used as immunogen, from genbank sequence NP_067032.3
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.