Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARD14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534363
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534363 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534363 Supplier Thermo Scientific Supplier No. PA534363
Add to Cart
Add to Cart

CARD14 Polyclonal Antibody

CARD14 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen CARD14
Applications Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CARD14
Gene Accession No. Q9BXL6
Gene Alias Bcl10-interacting MAGUK protein 2; BIMP2; Card14; CARD-containing MAGUK protein 2; Carma 2; CARMA2; caspase recruitment domain family member 14; caspase recruitment domain family, member 14; caspase recruitment domain-containing protein 14; PRP; PSORS2; PSS1
Gene Symbols CARD14
Host Species Rabbit
Immunogen A 14 amino acid synthetic peptide near the carboxy terminus of human CARMA2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79092
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.