Learn More
Invitrogen™ CARD14 Polyclonal Antibody

Description
The antibody detects endogenous levels of total CARD14 / CARMA2 protein.
Specifications
Specifications
| Antigen | CARD14 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CARD14 |
| Gene Accession No. | Q9BXL6 |
| Gene Alias | Bcl10-interacting MAGUK protein 2; BIMP2; Card14; CARD-containing MAGUK protein 2; Carma 2; CARMA2; caspase recruitment domain family member 14; caspase recruitment domain family, member 14; caspase recruitment domain-containing protein 14; PRP; PSORS2; PSS1 |
| Gene Symbols | CARD14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.