Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARD17 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987720
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7720 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7720 Supplier Bioss Supplier No. BS7090R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen CARD17
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CARD17
Gene Accession No. Q5XLA6
Gene Alias CARD17; caspase recruitment domain family member 17; caspase recruitment domain family, member 17; Caspase recruitment domain-containing protein 17; Caspase-1 inhibitor INCA; INCA; Inhibitory CARD; inhibitory caspase recruitment domain (CARD) protein; Inhibitory caspase recruitment domain protein
Gene Symbols CARD17
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CARD17/INCA.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440068
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.