Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARD6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987717
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7717 Supplier Bioss Supplier No. BS7087R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein that contains a caspase recruitment domain (CARD), an antiparallel six-helical bundle that mediates homotypic protein-protein interactions. The encoded protein is a microtubule-associated protein that has been shown to interact with receptor-interacting protein kinases and positively modulate signal transduction pathways converging on activation of the inducible transcription factor NF-kB.

Specifications

Antigen CARD6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CARD6
Gene Accession No. Q9BX69
Gene Alias CARD6; CARD-containing inhibitor of Nod1 and Cardiak-induced NF-kB activation; caspase recruitment domain family member 6; caspase recruitment domain family, member 6; caspase recruitment domain protein 6; Caspase recruitment domain-containing protein 6; CINCIN1; D730008L15
Gene Symbols CARD6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CARD6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84674
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.