Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARD9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP237975
Description
CARD9 Polyclonal specifically detects CARD9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CARD9 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Q9H257 | |
| CARD9 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GSPKQPFAALHQEQVLRNPHDAGLSSGEPPEKERRRLKESFENYRRKRALRKMQKGWRQGEEDRENTTGSDNTDTEGS | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CANDF2, caspase recruitment domain family, member 9, caspase recruitment domain-containing protein 9, hCARD9 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 64170 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction