Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CARD9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519993
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519993 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519993 Supplier Invitrogen™ Supplier No. PA519993
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In Western blot applications, this antibody detects a band at ~59kDa. A suggested positive control is PC-3 cell lysate. PA5-19993 can be used with blocking peptide PEP-0115.

The protein encoded by this gene is a member of the CARD protein family, which is defined by the presence of a characteristic caspase-associated recruitment domain (CARD). CARD is a protein interaction domain known to participate in activation or suppression of CARD containing members of the caspase family, and thus plays an important regulatory role in cell apoptosis. This protein was identified by its selective association with the CARD domain of BCL10, a postive regulator of apoptosis and NF-kappaB activation, and is thought to function as a molecular scaffold for the assembly of a BCL10 signaling complex that activates NF-kappaB. Several alternatively spliced transcript variants have been observed, but their full-length nature is not clearly defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARD9
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CARD9
Gene Accession No. Q9H257
Gene Alias CANDF2; card 9; Card9; caspase recruitment domain family member 9; caspase recruitment domain family, member 9; caspase recruitment domain protein 9; caspase recruitment domain-containing protein 9; Gm782; hCARD9; rCARD9; RP11-413M3.9
Gene Symbols CARD9
Host Species Rabbit
Immunogen A 16 amino acid peptide near the carboxy terminus of human CARD9. The immunogen islocated within the last 50 amino acidsof CARD9.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64170
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.