Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CARD9 Recombinant Rabbit Monoclonal Antibody (PSH0-41)
GREENER_CHOICE

Catalog No. FEMA550828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550828 Supplier Invitrogen™ Supplier No. MA550828
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: HL-60 cell lysate, THP-1 cell lysate, RAW264.7 cell lysate, PC-12 cell lysate, THP-1, PC-3. Predicted band size: 62.2 kDa Subcellular Location: Cytoplasm.

The protein encoded by this gene is a member of the CARD protein family, which is defined by the presence of a characteristic caspase-associated recruitment domain (CARD). CARD is a protein interaction domain known to participate in activation or suppression of CARD containing members of the caspase family, and thus plays an important regulatory role in cell apoptosis. This protein was identified by its selective association with the CARD domain of BCL10, a postive regulator of apoptosis and NF-kappaB activation, and is thought to function as a molecular scaffold for the assembly of a BCL10 signaling complex that activates NF-kappaB. Several alternatively spliced transcript variants have been observed, but their full-length nature is not clearly defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARD9
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone PSH0-41
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CARD9
Gene Accession No. A2AIV8, Q9EPY0, Q9H257
Gene Alias CANDF2; card 9; Card9; caspase recruitment domain family member 9; caspase recruitment domain family, member 9; caspase recruitment domain protein 9; caspase recruitment domain-containing protein 9; Gm782; hCARD9; rCARD9; RP11-413M3.9
Gene Symbols CARD9
Host Species Rabbit
Immunogen Recombinant protein within human CARD9 aa 1-200/536.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 332579, 64170, 64171
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.