Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Cardboard Cryogenic Freezer Box Partition/Divider
GREENER_CHOICE

Catalog No. 17100457
Change view
Click to view available options
Holds:
100 Cryogenic Vials
49 Cryogenic Vials
64 Cryogenic Vials
81 Cryogenic Vials
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
17-100-457 81 Cryogenic Vials
17-100-450 100 Cryogenic Vials
17-100-428 64 Cryogenic Vials
17-100-459 49 Cryogenic Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-100-457 Supplier Heathrow Scientific™ Supplier No. HS2860H
Add to Cart
Add to Cart

Box for storage of small samples and tubes.

  • Array: 10 x 10
  • Low Temp: -196°C

Specifications

Color White
Material Cardboard
Shape Square
Type Box Divider
For Use With (Equipment) 1.5/2.0 mL Cryogenic Vials
Holds 81 Cryogenic Vials
Tube Array 9 x 9
Withstand Temperature (Metric) -196°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.